Product Image
TriPlus RSH Autosampler Fixed GT Needle Syringe
Thermo Scientific™ TriPlus RSH Autosampler Fixed GT Needle Syringes are accurate and reproducible over a range of volumes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Needle Gauge
365H2181100 μL23 G
365K28111 mL22 G
365D033110 μL26s G
365H2161100 μL26s G
365I2351250 μL23 G
Catalog number 365H2181
Price (USD)
-
Volume (Metric):
100 μL
Needle Gauge:
23 G
Thermo Scientific™ TriPlus RSH Autosampler Fixed GT Needle Syringes are accurate and reproducible over a range of volumes.
  • Borosilicate glass with metal flanges
  • Needles and plungers are stainless steel
  • Field-proven syringe design
Specifications
Description100 μL FN GT, 23 G, 57 mm
Length (Metric)57 mm
Length (Metric) Needle57 mm
Needle Point StyleSide Hole
PTFE Tipped PlungerYes
Product TypeGC Autosampler Syringe
QuantityEa.
Connection TypeFixed
For Use With (Equipment)TriPlus RSH Autosampler
MaterialStainless Steel (Needle), Stainless Steel (Plunger)
Needle Gauge23 G
Product LineTriPlus
Volume (Metric)100 μL
Unit SizeEach