Syringes for Thermo Scientific™ HPLC Instruments
Thermo Scientific™
Syringes for Thermo Scientific™ HPLC Instruments
Thermo Scientific™ Syringes for Thermo Scientific HPLC Instruments offer easy, accurate and reproducible injections.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)For Use With (Equipment)
365ILT91250 μLAS1000, AS3000 Autosampler
365ILT21250 μLLCQ HPLC
365JLT41500 μLLCQ HPLC
365JLT61500 μLAS1000, AS3000 Autosampler
365LLT812.5 mLAS3000, AS3500 Autosampler
Catalog number 365ILT91
Price (USD)
268.08
Each
Volume (Metric):
250 μL
For Use With (Equipment):
AS1000, AS3000 Autosampler
Price (USD)
268.08
Each
Thermo Scientific™ Syringes for Thermo Scientific HPLC Instruments offer easy, accurate and reproducible injections.
  • Square tip to prevent damage to the injector
  • Wide range of volumes
  • Precision made from borosilicate glass and stainless steel
  • Robust design and easy-to-read markings
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DisposableReusable
Product TypeHPLC Autosampler Syringe
For Use With (Equipment)AS1000, AS3000 Autosampler
Volume (Metric)250 μL
Unit SizeEach