Product Image
Gibco™

McCoy's 5A (Modified) Medium, GlutaMAX™ Supplement

McCoy's 5A (modified) is a general purpose medium that supports the propagation of many types of primary cells, established cellLeia mais
Have Questions?
Número do catálogoQuantity
36600088
conhecido também como 36600-088
10 x 500 mL
Número do catálogo 36600088
conhecido também como 36600-088
Preço (BRL)
-
Quantity:
10 x 500 mL
Customize this product

McCoy's 5A (modified) is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues. This medium will support the growth of primary mammalian cells derived from normal bone marrow, skin, spleen, kidney, lung, rat embryos, and other tissues.


This McCoy's 5A is modified as follows:
WithWithout
• High Glucose• Sodium Pyruvate
• GlutaMAX™• HEPES
• Bacto-peptone 
• Phenol Red 


The complete formulation is available.

Dr. Thomas McCoy originally formulated McCoy's 5A medium as a modification of Basal Medium 5A. Unlike other media, McCoy's 5A contains the reducing agent glutathione, bacto-peptone, and a high level of glucose. This product also includes Dr. Hsu's addition of Hanks' salts to enable use outside a CO2 incubator.

Gibco™ McCoy's 5A (modified) requires serum supplementation, commonly with 10% Fetal Bovine Serum (FBS). Gibco™ McCoy's 5A (modified) uses a sodium bicarbonate buffer system (2.2 g / L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Especificações
Cell TypeMammalian Cells
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeMcCoy's 5A Medium
Quantity10 x 500 mL
Shelf Life12 Months
Culture TypeMammalian Cell Culture
FormLiquid
SterilitySterile-filtered
With AdditivesHigh Glucose, Glutamine, Phenol Red, Sodium Pyruvate
Without AdditivesNo HEPES
Unit SizeEach

Frequently asked questions (FAQs)

Will depletion, absence, or breakdown of essential growth-promoting components such as glutamine or growth factors reduce the growth rate of my culture?

Yes. If you suspect that this is the case, remove the medium and add fresh medium. Alternatively, you can supplement medium with growth-promoting components. It is also possible to substitute GlutaMax I or II for glutamine in the medium to prevent glutamine exhaustion.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Will the media formulations containing GlutaMAX supplement change with respect to L-glutamine content?

In all media containing GlutaMAX supplement dipeptides as a substitute for L-glutamine, concentration is equimolar with the L-glutamine in the original formulation.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.