4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 97%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 97%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 97%
Thermo Scientific Chemicals

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 97%

CAS: 180516-87-4 | C13H17BO4 | 248.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 366390010
Price (USD)
32.65
Online exclusive
36.40
Save 3.75
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
32.65
Online exclusive
36.40
Save 3.75
Each
Chemical Identifiers
CAS180516-87-4
IUPAC Name4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Molecular FormulaC13H17BO4
InChI KeyIYDKBQIEOBXLTP-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Appearance (Form)Crystalline powder or crystals or needles
Infrared spectrumConforms
Melting point227°C to 232°C
Titration with NaOH96.0 to 104.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only