4-Hydroxypiperidine, 99+%
4-Hydroxypiperidine, 99+%
4-Hydroxypiperidine, 99+%
Thermo Scientific Chemicals

4-Hydroxypiperidine, 99+%

CAS: 5382-16-1 | C5H11NO | 101.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 366510250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS5382-16-1
IUPAC Name4-hydroxypiperidin-1-ium
Molecular FormulaC5H12NO
InChI KeyHDOWRFHMPULYOA-UHFFFAOYSA-O
SMILESOC1CC[NH2+]CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<1 % (K.F.)
GC>=99.0 %
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only