2-(2-Thienyl)ethanol, 98%
2-(2-Thienyl)ethanol, 98%
2-(2-Thienyl)ethanol, 98%
Thermo Scientific Chemicals

2-(2-Thienyl)ethanol, 98%

CAS: 5402-55-1 | C6H8OS | 128.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
366541000100 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 366541000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5402-55-1
IUPAC Name2-(thiophen-2-yl)ethan-1-ol
Molecular FormulaC6H8OS
InChI KeyVMJOFTHFJMLIKL-UHFFFAOYSA-N
SMILESOCCC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay darken on storage
Infrared spectrumConforms
Refractive index1.5490 to 1.5520 (20°C, 589 nm)
GC>=97.5 %
Appearance (Color)Clear light yellow to grey-green to brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only