Search
View additional product information for Nalgene™ Fluorocarbon (PFA) Resin Plastic Graduated Cylinder - FAQs (3666-0100)
1 product FAQs found
We do not offer plastic graduated cylinders with Class A accuracy. It is very difficult to reach Class A accuracy with plastic, which is why they are almost always glass.