Tri-tert-butylphosphonium tetrafluoroborate, 97+%
Tri-tert-butylphosphonium tetrafluoroborate, 97+%
Tri-tert-butylphosphonium tetrafluoroborate, 97+%
Thermo Scientific Chemicals

Tri-tert-butylphosphonium tetrafluoroborate, 97+%

CAS: 131274-22-1 | C12H28BF4P | 290.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 366940010
Price (USD)
84.65
Special offer
Online exclusive
Ends: 28-Dec-2026
99.00
Save 14.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
84.65
Special offer
Online exclusive
Ends: 28-Dec-2026
99.00
Save 14.35
Each
Chemical Identifiers
CAS131274-22-1
IUPAC Nametetrafluoroboranuide; tri-tert-butylphosphanium
Molecular FormulaC12H28BF4P
InChI KeyYTJUCJAUJCXFTN-UHFFFAOYSA-O
SMILESF[B-](F)(F)F.CC(C)(C)[PH+](C(C)(C)C)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals and/or chunks
Infrared spectrumConforms
NMR (31P-NMR)>=97.0 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only