Calcium thioglycolate trihydrate, 99%
Calcium thioglycolate trihydrate, 99%
Calcium thioglycolate trihydrate, 99%
Thermo Scientific Chemicals

Calcium thioglycolate trihydrate, 99%

CAS: 65208-41-5 | C2H10CaO5S | 186.24 g/mol
Have Questions?
Catalog NumberQuantityPackaging
367035000500 gPlastic bottle
Catalog number 367035000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS65208-41-5
IUPAC Name2-sulfanylacetic acid calcium trihydrate
Molecular FormulaC2H10CaO5S
InChI KeyGDECCIJZCWLFSB-UHFFFAOYSA-N
SMILESO.O.O.[Ca].OC(=O)CS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
Titration Iodometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only