Benzeneruthenium(II) chloride, dimer, 97%
Benzeneruthenium(II) chloride, dimer, 97%
Benzeneruthenium(II) chloride, dimer, 97%
Thermo Scientific Chemicals

Benzeneruthenium(II) chloride, dimer, 97%

CAS: 37366-09-9 | C12H12Cl4Ru2 | 500.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 367480050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS37366-09-9
IUPAC Namebis(benzene); bis(dichlororuthenium)
Molecular FormulaC12H12Cl4Ru2
InChI KeyYGXMUPKIEHNBNQ-UHFFFAOYSA-J
SMILESCl[Ru]Cl.Cl[Ru]Cl.C1=CC=CC=C1.C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to brown
Infrared spectrumConforms
Ruthenium (Ru)39.2 to 41.0 %
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only