cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate, 97%
cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate, 97%
cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate, 97%
Thermo Scientific Chemicals

cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate, 97%

CAS: 98014-14-3 | C20H16Cl2N4Ru | 484.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number 367500010
Price (EUR)
275,00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
275,00
Each
Chemical Identifiers
CAS98014-14-3
IUPAC Namebis(2,2'-bipyridine); dichlororuthenium
Molecular FormulaC20H16Cl2N4Ru
InChI KeyMGAJEYXQYJBLQL-UHFFFAOYSA-L
SMILESCl[Ru]Cl.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Assay>=96.0 % (Based on Carbon)
Appearance (Color)Dark green to black
Appearance (Form)Powder and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only