3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 97%, Thermo Scientific Chemicals
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 97%, Thermo Scientific Chemicals
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 97%, Thermo Scientific Chemicals

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 97%, Thermo Scientific Chemicals

CAS: 269409-73-6 | Molecular Formula: C13H17BO4 | Molecular Weight: 248.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3675700505 gGlass bottle
Catalog number 367570050
Price (USD)
135.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
135.00
Each
Chemical Identifiers
CAS269409-73-6
IUPAC Name3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Molecular FormulaC13H17BO4
InChI KeyOPWAPCOSDAFWFB-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC=CC(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=96.0 %
Melting point206°C to 211°C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only