(Bromomethyl)cyclobutane, 99%
(Bromomethyl)cyclobutane, 99%
(Bromomethyl)cyclobutane, 99%
Thermo Scientific Chemicals

(Bromomethyl)cyclobutane, 99%

CAS: 17247-58-4 | C5H9Br | 149.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 367640250
Price (USD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS17247-58-4
IUPAC Name(bromomethyl)cyclobutane
Molecular FormulaC5H9Br
InChI KeyFLHFTXCMKFVKRP-UHFFFAOYSA-N
SMILESBrCC1CCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4790 to 1.4830 (20°C, 589 nm)
GC>=98.5 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only