3-Amino-5-methylpyrazole, 97%
3-Amino-5-methylpyrazole, 97%
3-Amino-5-methylpyrazole, 97%
Thermo Scientific Chemicals

3-Amino-5-methylpyrazole, 97%

CAS: 31230-17-8 | C4H7N3 | 97.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 368030250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS31230-17-8
IUPAC Name5-methyl-1H-pyrazol-3-amine
Molecular FormulaC4H7N3
InChI KeyFYTLHYRDGXRYEY-UHFFFAOYSA-N
SMILESCC1=CC(N)=NN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to orange
Appearance (Form)Solid or viscous liquid
GC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only