N-BOC-4-Hydroxypiperidine, 97%
N-BOC-4-Hydroxypiperidine, 97%
N-BOC-4-Hydroxypiperidine, 97%
N-BOC-4-Hydroxypiperidine, 97%
Thermo Scientific Chemicals

N-BOC-4-Hydroxypiperidine, 97%

CAS: 109384-19-2 | C10H19NO3 | 201.27 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
產品號碼 368270250
價格 (TWD)
-
申請報價
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS109384-19-2
IUPAC Nametert-butyl 4-hydroxypiperidine-1-carboxylate
Molecular FormulaC10H19NO3
InChI KeyPWQLFIKTGRINFF-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CCC(O)CC1
檢視更多
規格Specification Sheet規格表
Appearance (Form)Powder
Appearance (Color)White to cream
GC>=96.0 %
Melting point60°C to 65°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only