Benzophenone imine, 97%, stabilized
Benzophenone imine, 97%, stabilized
Benzophenone imine, 97%, stabilized
Thermo Scientific Chemicals

Benzophenone imine, 97%, stabilized

CAS: 1013-88-3 | C13H11N | 181.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 368591000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1013-88-3
IUPAC Namediphenylmethanimine
Molecular FormulaC13H11N
InChI KeySXZIXHOMFPUIRK-UHFFFAOYSA-N
SMILESN=C(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow-brown
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.6170 to 1.6190 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only