(±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98%
(±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98%
(±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98%
Thermo Scientific Chemicals

(±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98%

CAS: 98327-87-8 | C44H32P2 | 622.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number 368640250
Price (USD)
266.65
Special offer
Online exclusive
Ends: 28-Dec-2026
314.00
Save 47.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
266.65
Special offer
Online exclusive
Ends: 28-Dec-2026
314.00
Save 47.35
Each
Chemical Identifiers
CAS98327-87-8
IUPAC Name[2'-(diphenylphosphanyl)-[1,1'-binaphthalen]-2-yl]diphenylphosphane
Molecular FormulaC44H32P2
InChI KeyMUALRAIOVNYAIW-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)P(C1=CC=CC=C1)C1=C(C2=CC=CC=C2C=C1)C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Infrared spectrumConforms
Appearance (Color)White to light beige
Appearance (Form)Powder
Melting point280°C to 285°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only