Oxalyl chloride, 2.0M solution in dichloromethane, AcroSeal™
Oxalyl chloride, 2.0M solution in dichloromethane, AcroSeal™
Oxalyl chloride, 2.0M solution in dichloromethane, AcroSeal™
Thermo Scientific Chemicals

Oxalyl chloride, 2.0M solution in dichloromethane, AcroSeal™

CAS: 79-37-8 | C2Cl2O2 | 126.93 g/mol
Have Questions?
Catalog NumberQuantityPackaging
369181000100 mLAcroSeal™ Glass bottle
Catalog number 369181000
Price (USD)
67.60
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)
67.60
Each
Chemical Identifiers
CAS79-37-8
IUPAC Nameoxalic dichloride
Molecular FormulaC2Cl2O2
InChI KeyCTSLXHKWHWQRSH-UHFFFAOYSA-N
SMILESClC(=O)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Titration Argentometric1.9 to 2.1 M
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Oxidation reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only