Tris(dibenzylideneacetone)dipalladium-chloroform adduct, 97%
Tris(dibenzylideneacetone)dipalladium-chloroform adduct, 97%
Tris(dibenzylideneacetone)dipalladium-chloroform adduct, 97%
Thermo Scientific Chemicals

Tris(dibenzylideneacetone)dipalladium-chloroform adduct, 97%

CAS: 52522-40-4 | C52H43Cl3O3Pd2 | 1035.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 369340010
Price (USD)
107.65
Online exclusive
119.00
Save 11.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
107.65
Online exclusive
119.00
Save 11.35
Each
Chemical Identifiers
CAS52522-40-4
IUPAC Nametris(1,5-diphenylpenta-1,4-dien-3-one) trichloromethane dipalladium
Molecular FormulaC52H43Cl3O3Pd2
InChI KeyLNAMMBFJMYMQTO-UHFFFAOYSA-N
SMILES[Pd].[Pd].ClC(Cl)Cl.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=96.0 % (ex Pd)
Appearance (Color)Purple to black
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only