Chlorobis(cyclooctene)rhodium(I) dimer, 98%
Chlorobis(cyclooctene)rhodium(I) dimer, 98%
Chlorobis(cyclooctene)rhodium(I) dimer, 98%
Thermo Scientific Chemicals

Chlorobis(cyclooctene)rhodium(I) dimer, 98%

CAS: 12279-09-3 | C32H56Cl2Rh2 | 717.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Catalog number 369371000
Price (USD)
-
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS12279-09-3
IUPAC Namebis(λ¹-rhodium(1+)) tetrakis((Z)-cyclooctene) dichloride
Molecular FormulaC32H56Cl2Rh2
InChI KeyGQPAPAIPOLEZHT-XFCUKONHSA-L
SMILES[Cl-].[Cl-].[Rh+].[Rh+].C1CCC\C=C/CC1.C1CCC\C=C/CC1.C1CCC\C=C/CC1.C1CCC\C=C/CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Rhodium (Rh)28.0 to 29.4 %
Infrared spectrumConforms
Appearance (Color)Orange-brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only