4-Hydroxyphenylboronic acid, 97%
4-Hydroxyphenylboronic acid, 97%
4-Hydroxyphenylboronic acid, 97%
Thermo Scientific Chemicals

4-Hydroxyphenylboronic acid, 97%

CAS: 71597-85-8 | C6H7BO3 | 137.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 369420050
Price (USD)
113.65
Special offer
Online exclusive
Ends: 28-Dec-2026
134.00
Save 20.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
113.65
Special offer
Online exclusive
Ends: 28-Dec-2026
134.00
Save 20.35
Each
Chemical Identifiers
CAS71597-85-8
IUPAC Name(4-hydroxyphenyl)boronic acid
Molecular FormulaC6H7BO3
InChI KeyCOIQUVGFTILYGA-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light brown
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only