1-Decanesulfonic acid, sodium salt, 96%
1-Decanesulfonic acid, sodium salt, 96%
1-Decanesulfonic acid, sodium salt, 96%
Thermo Scientific Chemicals

1-Decanesulfonic acid, sodium salt, 96%

1-Decanesulfonic acid, sodium salt, 96%, Quantity: 25g, Packaging: Glass bottle, White to Cream, Melting Point: >300 deg.C, Molecular Weight: 244.32, Percent Purity: 96%, Assay Percent Range: 96%, Beilstein: 04, III, 27, CAS: 13419-61-9, CAS Max %: 100.0, CAS Min %: 95.0 | CAS: 13419-61-9 | C10H21NaO3S | 244.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 369450050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13419-61-9
IUPAC Namesodium decane-1-sulfonate
Molecular FormulaC10H21NaO3S
InChI KeyAIMUHNZKNFEZSN-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<5 % (100°C, 3 h) (vacuum)
Titration after Ion exchange>=95 % (On dry substance)
Appearance (Color)White to cream
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only