6-Aminoquinoline, 98%
6-Aminoquinoline, 98%
6-Aminoquinoline, 98%
Thermo Scientific Chemicals

6-Aminoquinoline, 98%

CAS: 580-15-4 | C9H8N2 | 144.18 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
產品號碼 369640250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS580-15-4
IUPAC Namequinolin-6-amine
Molecular FormulaC9H8N2
InChI KeyRJSRSRITMWVIQT-UHFFFAOYSA-N
SMILESNC1=CC=C2N=CC=CC2=C1
檢視更多
規格Specification Sheet規格表
Titration with HClO497.5 to 102.5 %
Appearance (Form)Powder
Melting point115°C to 119°C
Appearance (Color)Yellow to grey-green to brown
Infrared spectrumConforms
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only