CryoBoxes, 81, 3.6mL cryotube, white - FAQs
View additional product information for CryoBoxes™ - FAQs (374187PK, 340053, 341483, 5025-0505, 374187, 867013-0240, 378247, 867013-0241, 330821, 867013-0244, 867013-0245, 341483PK, 867013-0242, 867013-0243, 5026-0909, 369647, 5027-0909, 340061, 5025-0505PK, 330821PK, 5026-0909PK, 5027-0909PK, 378220, 369639, 5026-1010)
1 product FAQs found
Can I purchase the dividers for either the 25 cell or 81 cell chipboard Nunc Cryoboxes separately?
The dividers are supplied with the chipboard Nunc Cryoboxes and not sold separately.