Bis(pyridine)iodonium tetrafluoroborate, 97%
Bis(pyridine)iodonium tetrafluoroborate, 97%
Bis(pyridine)iodonium tetrafluoroborate, 97%
Thermo Scientific Chemicals

Bis(pyridine)iodonium tetrafluoroborate, 97%

CAS: 15656-28-7 | C10H10BF4IN2 | 371.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 369680010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS15656-28-7
IUPAC Namebis(pyridin-1-ium-1-yl)iodanuide; tetrafluoroboranuide
Molecular FormulaC10H10BF4IN2
InChI KeyBMDSRCBKJZCUBH-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.[I-]([N+]1=CC=CC=C1)[N+]1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Titration with HClO4>=96.0 %
Appearance (Color)Light yellow to orange to tan
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only