(R)-(+)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene
(R)-(+)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene
(R)-(+)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene
Thermo Scientific Chemicals

(R)-(+)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene

CAS: 112022-83-0 | C18H20BNO | 277.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 369780050
Price (USD)
83.65
Online exclusive
92.80
Save 9.15
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
83.65
Online exclusive
92.80
Save 9.15
Each
Chemical Identifiers
CAS112022-83-0
IUPAC Name(3aR)-1-methyl-3,3-diphenyl-hexahydropyrrolo[1,2-c][1,3,2]oxazaborole
Molecular FormulaC18H20BNO
InChI KeyVMKAFJQFKBASMU-QGZVFWFLSA-N
SMILES[H][C@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Colorless to amber
Strength0.90 to 1.10 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only