Eaton's reagent
Eaton's reagent
Eaton's reagent
Eaton's reagent
Thermo Scientific Chemicals

Eaton's reagent

CAS: 39394-84-8 | O10P4
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Catalog number 369805000
Price (USD)
174.00
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
174.00
Each
Chemical Identifiers
CAS39394-84-8
IUPAC Namediphosphooxidane; methanesulfonic acid
Molecular FormulaCH4O8P2S
InChI KeyJHNLZOVBAQWGQU-UHFFFAOYSA-N
SMILESCS(O)(=O)=O.O=P(=O)OP(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)to light brown or pale pink
Appearance (Form)Solution
Titration with NaOH6.5 to 10.0 % (P2O5)
Appearance (Color)Clear colorless to yellow to orange
AppearanceMay contain precipitate, re-dissolves on warming
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only