Search
| Catalog Number | Overall Length (Metric) | Diameter (English) Inner | Color |
|---|---|---|---|
| 37005-50 | 50 cm | 0.005 in. | Red |
| 37007-100 | 100 cm | 0.007 in. | Yellow |
| 37010-100 | 100 cm | 0.010 in. | Blue |
Use Thermo Scientific™ Rheodyne™ Sample Loops for your Rheodyne HPLC sample injectors. They are available in stainless steel or biocompatible PEEK.