Sodium, dry stick
Sodium, dry stick
Sodium, dry stick
Thermo Scientific Chemicals

Sodium, dry stick

CAS: 7440-23-5 | Na | 22.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 370212500
Price (EUR)
148,00
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
148,00
Each
Chemical Identifiers
CAS7440-23-5
IUPAC Namesodium
Molecular FormulaNa
InChI KeyKEAYESYHFKHZAL-UHFFFAOYSA-N
SMILES[Na]
View more
SpecificationsSpecification SheetSpecification Sheet
Potassium (K)=<300 ppm
Appearance (Form)Sticks
Calcium (Ca)=<400 ppm
Appearance (Color)Light grey or silver white
Sodium (Na)>=99.8 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only