Due to high demand, order processing and delivery times for non-filtered tips may be longer than usual. Compare options for filtered tips »
SoftFit-L™ Empty Racks for use with SoftFit-L Reload Inserts and Reload System Towers
SoftFit-L™ Empty Racks for use with SoftFit-L Reload Inserts and Reload System Towers
Thermo Scientific™ART SoftFit-L™ empty racks, when used in conjunction with SoftFit-L reload towers and inserts, offer an environmentally friendly packaging option with less plastic waste than traditional racks. Compatible with ART SoftFit-L reload inserts and reload towers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Quantity
3704-ER20 μL Pipet Tip, 200 μL Pipet Tip, 300 μL Pipet Tip10 Racks/Pk., 5 Pks./Cs.
3706-ERRainin LTS LiteTouch8 Racks/Pk., 4 Pks./Cs.
Catalog number 3704-ER
Price (USD)
-
For Use With (Equipment):
20 μL Pipet Tip, 200 μL Pipet Tip, 300 μL Pipet Tip
Quantity:
10 Racks/Pk., 5 Pks./Cs.
Thermo Scientific™ART SoftFit-L™ empty racks, when used in conjunction with SoftFit-L reload towers and inserts, offer an environmentally friendly packaging option with less plastic waste than traditional racks. Compatible with ART SoftFit-L reload inserts and reload towers.
Specifications
ModelSoftFit L Rack
Product TypePipet Tip Rack
Quantity10 Racks/Pk., 5 Pks./Cs.
AutoclavableNot Autoclavable
For Use With (Equipment)20 μL Pipet Tip, 200 μL Pipet Tip, 300 μL Pipet Tip
Product LineART SoftFit
Unit SizeCase of 50