(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine, 97+%, Thermo Scientific Chemicals
(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine, 97+%, Thermo Scientific Chemicals
(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 370671000
Price (USD)
-
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS155806-35-2
Molecular FormulaC38H50FeOP2
InChI KeyYOYYLEHTACDQJL-BHACGSQPNA-N
SMILES[Fe].CCO.c1cccc1.C[C@@H](P(C1CCCCC1)C1CCCCC1)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)640.61
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only