SuperSignal™ West Dura Extended Duration Substrate
Thermo Scientific™

SuperSignal™ West Dura Extended Duration Substrate

Luminol-based enhanced chemiluminescence (ECL) horseradish peroxidase (HRP) substrate ideal for quantitative western blotting with stable light output for mid-femtogram level detection for western blot analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
3707120 mL
34075100 mL
34075X44 x 100 mL
34076200 mL
34076X44 x 200 mL
Catalog number 37071
Price (USD)
129.65
Online exclusive
149.00
Save 19.35
Each
Quantity:
20 mL
Request bulk or custom format
Price (USD)
129.65
Online exclusive
149.00
Save 19.35
Each

Thermo Scientific SuperSignal West Dura Extended Duration Substrate is a luminol-based enhanced chemiluminescence (ECL) horseradish peroxidase (HRP) substrate ideal for quantitative western blotting with stable light output for mid-femtogram level detection for western blot analysis.

SuperSignal West Dura Extended Duration Substrate characteristics include:

  • Sensitivity: mid-femtogram
  • Stability: 24-hr working solution stability; 1-year kit stability at room temperature
  • Compatibility: nitrocellulose and PVDF membranes
  • Signal duration: 24 hours
  • Recommended primary antibody concentration: 1:1,000–1:50,000 dilution (0.02–1.0 μg/mL)
  • Recommended secondary antibody concentration: 1:50,000–1:250,000 dilution (4–20 ng/mL)

SuperSignal West Dura Extended Duration Substrate for HRP is optimized for high sensitivity and long signal duration, making it ideal for cooled CCD camera-based detection systems. Unlike substrates with signals that decline to barely detectable levels in 30–60 minutes, the signal produced with SuperSignal West Dura chemiluminescent substrate is stable for 24 hours, allowing multiple film or camera exposures.

For Research Use Only. Not for use in diagnostic procedures.

General References:

  1. Gangalum, R.K.et al.(2004).J. Biol. Chem. 279,43374-43377.
  2. MacInnis, B.L. and Campenot, R.B. (2004).Science 295,1536-1539.
  3. Sato, S.,et al.(2003).J. Biol. Chem. 278,15123-15127.
  4. Tokumaru, H.,et al.(2001).Cell 104,421-432.
Specifications
Recommended Antibody Concentrations2°: 1:50-250K (4-20ng/mL), 1°: 1:5K (0.02- 1μg/mL)
SensitivityMid-femtogram
Signal Duration24 hr.
Sufficient For2 Mini-Gel Blots, 200 sq. cm of Membrane
Detection MethodChemiluminescence
For Use With (Application)Western Blot
FormLiquid
Membrane CompatibilityNitrocellulose, PVDF
Product LineSuperSignal
Product TypeSubstrate
Quantity20 mL
SubstrateHorseradish Peroxidase
Unit SizeEach
Contents & Storage
Sufficient For: 2 mini-gel blots; 200 cm2 of membrane
• Luminol/Enhancer, 10 mL
• Stable Peroxide Buffer, 10 mL

Store at room temperature.

Frequently asked questions (FAQs)

What types of membrane work best with SuperSignal West Dura Extended Duration Substrate?

Most researchers use nitrocellulose or polyvinyldiflouride (PVDF) membranes with SuperSignal West Dura Extended Duration Substrate. Both work well, although nitrocellulose seems to be better suited in some applications than PVDF. In addition, charge-modified nylon membrane performs well with this substrate. Please also see our guide for choosing western blot membranes (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/western-blotting/transfer-proteins-western-blot/membranes-transfer-buffers-western-blotting/membranes-western-blotting.html).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Will SuperSignal West Dura Extended Duration Substrate work with nucleic acid blots (Southern blotting, etc.)?

Yes. However, SuperSignal West Dura Extended Duration Substrate was optimized for use in Western blots and is generally not sensitive enough for most nucleic acid protocols. For Southern and Northern blotting, use our North2South Chemiluminescent Nucleic Acid Hybridization and Detection Kit (Cat. No. 17097), North2South Chemiluminescent Substrate for HRP (Cat. No. 17295), or the Chemiluminescent Nucleic Acid Detection Kit, if probing for biotinylated probes (Cat. No. 89880).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

I used the SuperSignal West Dura Extended Duration Substrate. Why is the entire film black or showing a reversed image?

The antibody concentration (primary and secondary) is too high. Use the antibody dilutions described in the product instructions. Most background will disappear when the proper blocking reagent and a HRP conjugate concentration are used.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Why do I see more bands with the SuperSignal West Dura Extended Duration Substrate than I've seen in the past with other substrates?

Because this substrate is more sensitive than other chemiluminescent substrates you might detect low-abundance proteins that were not visible before. When using a more sensitive substrate, more careful optimization of blocking buffer steps and antibody concentrations is required.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What protocol should I use to strip and reprobe a blot detected with SuperSignal West Dura Extended Duration Substrate?

See Tech Tip # 23: Strip and reprobe Western blots.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.