tert-Butyl (R)-(+)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, 95%
tert-Butyl (R)-(+)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, 95%
tert-Butyl (R)-(+)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, 95%
Thermo Scientific Chemicals

tert-Butyl (R)-(+)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, 95%

CAS: 95715-87-0 | C11H19NO4 | 229.28 g/mol
Have Questions?
Catalog NumberQuantity
3708900101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 370890010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS95715-87-0
IUPAC Nametert-butyl (4R)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate
Molecular FormulaC11H19NO4
InChI KeyPNJXYVJNOCLJLJ-QMMMGPOBSA-N
SMILESCC(C)(C)OC(=O)N1[C@H](COC1(C)C)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4450 to 1.4490 (20°C, 589 nm)
Specific optical rotation+85° to +95° (20°C, 589 nm) (c=1, CHCl3)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Viscous liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only