tert-Butyl acrylate, 99%, stabilized
tert-Butyl acrylate, 99%, stabilized
tert-Butyl acrylate, 99%, stabilized
Thermo Scientific Chemicals

tert-Butyl acrylate, 99%, stabilized

CAS: 1663-39-4 | C7H12O2 | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
5 L
Packaging:
View all
Glass bottle
Catalog number 371130010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1663-39-4
IUPAC Nametert-butyl prop-2-enoate
Molecular FormulaC7H12O2
InChI KeyISXSCDLOGDJUNJ-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Refractive index1.4100 to 1.4120 (20°C, 589 nm)
Stabilizer10 to 20 ppm MEHQ
Color scale=<10 APHA
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only