Hydroxylapatite, for analysis
Hydroxylapatite, for analysis
Hydroxylapatite, for analysis
Thermo Scientific Chemicals

Hydroxylapatite, for analysis

CAS: 1306-06-5 | Ca5HO13P3 | 502.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
Packaging:
View all
Plastic Bottle
Catalog number 371260010
Price (USD)
151.65
Special offer
Online exclusive
Ends: 28-Dec-2026
178.00
Save 26.35
Each
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
151.65
Special offer
Online exclusive
Ends: 28-Dec-2026
178.00
Save 26.35
Each
Chemical Identifiers
CAS1306-06-5
IUPAC Namepentacalcium hydroxide triphosphate
Molecular FormulaCa5HO13P3
InChI KeyXYJRXVWERLGGKC-UHFFFAOYSA-D
SMILES[OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on ignition=<8 % (800°C)
Heavy metals (as Pb)=<10 ppm
Sulfate (SO4)=<5000 ppm
Iron (Fe)=<400 ppm
Fluoride (F)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only