Hydroxylapatite, for analysis
Hydroxylapatite, for analysis
Hydroxylapatite, for analysis
Thermo Scientific Chemicals

Hydroxylapatite, for analysis

CAS: 1306-06-5 | Ca5HO13P3 | 502.31 g/mol
数量:
25 g
100 g
1 kg
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 371261000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1306-06-5
IUPAC Namepentacalcium hydroxide triphosphate
Molecular FormulaCa5HO13P3
InChI KeyXYJRXVWERLGGKC-UHFFFAOYSA-D
SMILES[OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
さらに表示
Fluoride (F)=<50 ppm
Copper (Cu)=<20 ppm
Loss on ignition=<8 % (800°C)
Heavy metals (as Pb)=<10 ppm
Sulfate (SO4)=<5000 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only