1-Ethynyl-4-fluorobenzene, 98%
1-Ethynyl-4-fluorobenzene, 98%
1-Ethynyl-4-fluorobenzene, 98%
Thermo Scientific Chemicals

1-Ethynyl-4-fluorobenzene, 98%

CAS: 766-98-3 | C8H5F | 120.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 371370010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS766-98-3
IUPAC Name1-ethynyl-4-fluorobenzene
Molecular FormulaC8H5F
InChI KeyQXSWHQGIEKUBAS-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5155 to 1.5185 (20°C, 589 nm)
Appearance (Form)Liquid after melting
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only