Pinacolborane, 97%, stabilized
Pinacolborane, 97%, stabilized
Pinacolborane, 97%, stabilized
Thermo Scientific Chemicals

Pinacolborane, 97%, stabilized

CAS: 25015-63-8 | C6H13BO2 | 127.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 371630050
Price (USD)
54.65
Online exclusive
60.00
Save 5.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
54.65
Online exclusive
60.00
Save 5.35
Each
Chemical Identifiers
CAS25015-63-8
IUPAC Name4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular FormulaC6H13BO2
InChI KeyUCFSYHMCKWNKAH-UHFFFAOYSA-N
SMILESCC1(C)OBOC1(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Stabilizer0.1 to 1.5 % Triethylamine
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.3950 to 1.4000 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only