Due to high demand, order processing and delivery times for non-filtered tips may be longer than usual. Compare options for filtered tips »
SoftFit-L™ Pipette Tip Reload™ System
SoftFit-L™ Pipette Tip Reload™ System
SoftFit-L™ Pipette Tip Reload™ System
SoftFit-L™ Pipette Tip Reload™ System
SoftFit-L™ Pipette Tip Reload™ System

SoftFit-L™ Pipette Tip Reload™ System

Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityUnit Size
3722-RI20 μLNon-sterileCase of 4800
3722-RIPK
also known as 3722-RI
20 μLNon-sterilePack of 960
3732-RI300 μLNon-sterileCase of 4800
3732-RIPK
also known as 3732-RI
300 μLNon-sterilePack of 960
3742-RI1200 μLNon-sterileCase of 3072
3742-RIPK
also known as 3742-RI
1200 μLNon-sterilePack of 768
3752-RI200 μLNon-sterileCase of 4800
3752-RIPK
also known as 3752-RI
200 μLNon-sterilePack of 960
3782-RI1000 μLNon-sterileCase of 3072
3782-RIPK
also known as 3782-RI
1000 μLNon-sterilePack of 768
3721-RI20 μLSterileCase of 4800
3721-RIPK
also known as 3721-RI
20 μLSterilePack of 960
3731-RI300 μLSterileCase of 4800
3731-RIPK
also known as 3731-RI
300 μLSterilePack of 960
3741-RI1200 μLSterileCase of 3072
3741-RIPK
also known as 3741-RI
1200 μLSterilePack of 768
3751-RI200 μLSterileCase of 4800
3751-RIPK
also known as 3751-RI
200 μLSterilePack of 960
3781-RI1000 μLSterileCase of 3072
3781-RIPK
also known as 3781-RI
1000 μLSterilePack of 768
Catalog number 3722-RI
Price (USD)
365.65
Online exclusive
664.83
Save 299.18
Case of 4800
Volume (Metric):
20 μL
Sterility:
Non-sterile
Unit Size:
Case of 4800
Price (USD)
365.65
Online exclusive
664.83
Save 299.18
Case of 4800
Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.

The SoftFit-L Reload System is a complete product offering of pipet tips compatible with LTS™ pipetters made by Rainin Instrument, LLC. Designed in response to customer demands, this unique system provides preloaded pipet tips in color-coded inserts by volume size with the option of an easy-to-use 'push down' reload pack. Choose sterile or nonsterile tips, in five volume sizes, with or without our proprietary low-retention technology.

  • Reload towers are an environmentally-friendly packaging option that reduces plastic waste and necessary storage space
  • Compatible with SoftFit-L empty racks
  • Color-coded inserts allows quick identification of tip volume
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, and Endotoxin Free
Description20L, Non-Filtered, Non-sterile, Refill
Length (English)1.95 in.
Length (Metric)4.86cm
Model20L
ColorNatural
Color CodeRed
FilteredNon-filtered
For Use With (Equipment)Rainin LTS LiteTouch
FormatRefill
Product LineART SoftFit L
SterilityNon-sterile
Tip StyleUniversal
Volume (Metric)20 μL
Unit SizeCase of 4800