Triisopropyl phosphite, 96%, Thermo Scientific Chemicals
Triisopropyl phosphite, 96%, Thermo Scientific Chemicals
Triisopropyl phosphite, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triisopropyl phosphite, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 372380025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS116-17-6
IUPAC Nametris(propan-2-yl) phosphite
Molecular FormulaC9H21O3P
InChI KeySJHCUXCOGGKFAI-UHFFFAOYSA-N
SMILESCC(C)OP(OC(C)C)OC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=95 %
Refractive index1.4095 to 1.4115 (20°C, 589 nm)
Color scale=<25 APHA
Infrared spectrumConforms
Appearance (Form)Clear liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only