Sodium hydroxide, 95.0-100.5%, pellets, FCC, NF, GMP, J.T.Baker™
Sodium hydroxide, 95.0-100.5%, pellets, FCC, NF, GMP, J.T.Baker™
Avantor™

Sodium hydroxide, 95.0-100.5%, pellets, FCC, NF, GMP, J.T.Baker™

This product is GMP Manufactured and meets N.F. and F.C.C. Requirements.자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
수량:
125 g
500 g
2.5 kg
12 kg
25 kg
80 lb.
110 lb.
220 lb.
카탈로그 번호 3728-R
제품 가격(KRW)
-
견적 요청하기
수량:
110 lb.
This product is GMP Manufactured and meets N.F. and F.C.C. Requirements.
사양
끓는점1390°C (1013 hPa)
CAS1310-73-2
화학물질 이름 또는 재질Sodium hydroxide
색상White
용융점323°C
분자식NaOH
포장Polyethylene drum, open head
동의어Caustic soda, Lye, Sodium hydrate, Soda lye
등급FCC/NF
분자량(g/mol)40
백분율 순도95.0-100.5%
수량110 lb.
Unit SizeEach