Matrix™ 1.0mL Screw-Top Tubes, rack, no cap, 2D - FAQs
Matrix™ 1.0mL Screw-Top Tubes, rack, no cap, 2D - FAQs
View additional product information for Matrix™ 1.0mL ScrewTop Tubes - FAQs (3740BLU, 3741PUR, 3740PUR, 3741BLU, 3741-WP, 3740YEL, 3741YEL, 3740WHI, 3741GRE, 3740GRE, 3740, 3742AMB, 3740TS, 3741, 3741AMB, 3742, 3741WHI, 3741RED, 3740RED)
1 product FAQs found
Do you offer Thermo Scientific storage tubes for storing light-sensitive samples?
We offer the following 0.5 mL and 1.0 mL Matrix 2D ScrewTop amber-colored resin storage tubes with red screw caps:
Cat. No.; Description; Quantity 3741AMB; 1.0 mL, Amber, 2D Barcoded, V-bottom, ScrewTop with caps, Sterile; 5 latch racks of 96/case 3742AMB; 1.0 mL, Amber, 2D Barcoded, V-bottom, ScrewTop without caps, Sterile; 5 latch racks of 96/case 3743AMB; 0.5 mL, Amber, 2D Barcoded, V-bottom, ScrewTop with caps, Sterile; 5 latch racks of 96/case 3745AMB; 0.5 mL, Amber, 2D Barcoded, V-bottom, ScrewTop without caps, Sterile; 5 latch racks of 96/case