Nunc™ Universal Tri-coded Tubes
Thermo Scientific™
Nunc™ Universal Tri-coded Tubes
Use the automation friendly Thermo Scientific™ Nunc™ Universal Tri-coded Tubes to securely store and track precious samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberThread StylePackagingClosure Color
374516RED-WP1DInternalBulk, 10 bags of 48 tubesRed
374VB1-WP1DExternal10 bags of 48Clear
374VR1-WP1DExternal10 racks of 48Clear
374516-WP1DInternal10 bags of 48Clear
374322-WP1DExternal10 bags of 24Clear
374516ORG-WP1DInternal10 bags of 48Orange
374516YEL-WP1DInternal10 bags of 48Yellow
374516GRN-WP1DInternal10 bags of 48Green
374516PUR-WP1DInternal10 bags of 48Purple
374516BLU-WP1DInternal10 bags of 48 tubesBlue
Catalog number 374516RED-WP1D
Price (USD)
-
Thread Style:
Internal
Packaging:
Bulk, 10 bags of 48 tubes
Closure Color:
Red
Use the automation friendly Thermo Scientific™ Nunc™ Universal Tri-coded Tubes to improve the efficiency and throughput of your sample storage workflow. Tubes are molded from medical grade resins and feature a permanent, high contrast 2D, 1D linear and human readable barcode. Tri-coded tubes come sterile and offered in both racked and bulk formats. The 2D screwtop tubes meet a range of manual and automated long term storage needs and can be used in combination with Thermo Scientific barcode readers, arrayers and capping equipment.
Scalable
  • Tubes and rack are ideal for both lower-throughput and high-throughput use
  • Feature graduations and white patch for manual marking
  • Can be used in traditional cryostorage boxes made for 10x10 arrays
  • External threads limit contamination
  • Automation-pickable in Universal Latch Rack
Innovative Universal Cap Design
  • Single-piece construction with molded-in gasket
  • Cannot be over-tightened manually or with automated capping tools
  • Thermoplastic elastomer gasket material is compatible with storage at room temperature down to vapor phase of liquid nitrogen
Secure Tracking
  • Permanently-bonded, unique 2D barcode laser-etched onto the base of tube to securely identify and track samples
  • 2D barcode readers instantly scans each tube's 2D barcode, registering the tube in any application or database, or integrates via several export options
  • Scan the tube to access sample information and identify the contents
  • Tubes featured with 1D (linear) barcode and human readable on the side that matches 2D barcode on the bottom of the tube
Stringent Quality Control
  • Every 2D barcoded storage tube is scanned to ensure readability
  • Barcodes are checked against database of previously-assigned codes to prevent duplicates
  • Every tube has a Sterility Assurance Level of 106 and is made with class VI medical grade resins
Specifications
Barcode1D/2D/HR codes
Closure ColorRed
PackagingBulk, 10 bags of 48 tubes
AutoclavableNot Autoclavable
Bottom ShapeFlat
Capacity (Metric)2 mL
GraduatedYes
MaterialPolypropylene
Product TypeCryogenic Storage Tube
SterilitySterile
Thread StyleInternal
Unit SizeCase of 480