N-Phenylbis(trifluoromethanesulfonimide), 97%
N-Phenylbis(trifluoromethanesulfonimide), 97%
N-Phenylbis(trifluoromethanesulfonimide), 97%
Thermo Scientific Chemicals

N-Phenylbis(trifluoromethanesulfonimide), 97%

CAS: 37595-74-7 | C8H5F6NO4S2 | 357.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 374790050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS37595-74-7
IUPAC Name1,1,1-trifluoro-N-phenyl-N-trifluoromethanesulfonylmethanesulfonamide
Molecular FormulaC8H5F6NO4S2
InChI KeyDIOHEXPTUTVCNX-UHFFFAOYSA-N
SMILESFC(F)(F)S(=O)(=O)N(C1=CC=CC=C1)S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point99°C to 105°C
HPLC>=96.0 %
Appearance (Color)White to yellow to beige
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only