Mono-methyl phthalate, 98%
Mono-methyl phthalate, 98%
Mono-methyl phthalate, 98%
Thermo Scientific Chemicals

Mono-methyl phthalate, 98%

CAS: 4376-18-5 | C9H8O4 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 374880250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4376-18-5
IUPAC Name2-(methoxycarbonyl)benzoic acid
Molecular FormulaC9H8O4
InChI KeyFNJSWIPFHMKRAT-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point81°C to 86°C
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only