Due to high demand, order processing and delivery times for non-filtered tips may be longer than usual. Compare options for filtered tips »
SoftFit-L™ Non-Filtered Low Retention Pipette Tips in Racks with Lift-off Lid

SoftFit-L™ Non-Filtered Low Retention Pipette Tips in Racks with Lift-off Lid

Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Sterility
3752-05200 μLNon-sterile
3721-0520 μLSterile
3722-0520 μLNon-sterile
3751-05200 μLSterile
Catalog number 3752-05
Price (USD)
-
Volume (Metric):
200 μL
Sterility:
Non-sterile
Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
  • Low-retention tips maximize sample delivery
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, and Endotoxin Free
Description200L, Non-Filtered, Non-sterile, Rack
Length (English)3.42 in.
Length (Metric)8.67cm
Model200L
AutoclavableAutoclavable
ColorNatural
Color CodeYellow
FilteredNon-filtered
For Use With (Equipment)Rainin LTS LiteTouch
FormatRack
Product LineART SoftFit L
SterilityNon-sterile
Tip StyleUniversal
Volume (Metric)200 μL
Unit SizeCase of 4800