tert-Butyl methyl ether, 99+%, Extra Dry, AcroSeal™
tert-Butyl methyl ether, 99+%, Extra Dry, AcroSeal™
tert-Butyl methyl ether, 99+%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals

tert-Butyl methyl ether, 99+%, Extra Dry, AcroSeal™

CAS: 1634-04-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number 375210010
Price (USD)
102.65
Online exclusive
114.00
Save 11.35
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
102.65
Online exclusive
114.00
Save 11.35
Each
Chemical Identifiers
CAS1634-04-4
IUPAC Name2-methoxy-2-methylpropane
Molecular FormulaC5H12O
InChI KeyBZLVMXJERCGZMT-UHFFFAOYSA-N
SMILESCOC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %
Water=<0.005 %
Peroxides=<0.005 % (as H2O2)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only