Search
View additional product information for Fish Serum Blocking Buffer - FAQs (37527X3, 37527)
1 product FAQs found
This information is proprietary. The protein concentration in the Fish Serum Blocking Buffer has been optimized for direct usage without further dilution, however if desired, the blocking buffer may be diluted with phosphate-buffered saline. For best results, our recommendation is to empirically determine the optimal concentration to use.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.