Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Thermo Scientific™
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Store samples from general cold storage to the vapor phase of liquid nitrogen with these biobanking and cell culture cryogenic tubes, available with either internal or external threads.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure ColorBottom Shape
375353ORG1 mLOrangeConical (Starfoot)
337516RED4.5 mLRedRound (Starfoot)
337516ORG4.5 mLOrangeRound (Starfoot)
337516YEL4.5 mLYellowRound (Starfoot)
337516GRN4.5 mLGreenRound (Starfoot)
337516BLU4.5 mLBlueRound (Starfoot)
337516PUR4.5 mLPurpleRound (Starfoot)
375353RED1 mLRedConical (Starfoot)
375353YEL1 mLYellowConical (Starfoot)
375353GRN1 mLGreenConical (Starfoot)
375353BLU1 mLBlueConical (Starfoot)
375353PUR1 mLPurpleConical (Starfoot)
375418RED1.8 mLRedRound (Starfoot)
375418ORG1.8 mLOrangeRound (Starfoot)
375418YEL1.8 mLYellowRound (Starfoot)
375418GRN1.8 mLGreenRound (Starfoot)
375418BLU1.8 mLBlueRound (Starfoot)
375418PUR1.8 mLPurpleRound (Starfoot)
Catalog number 375353ORG
Price (USD)
-
Capacity (Metric):
1 mL
Closure Color:
Orange
Bottom Shape:
Conical (Starfoot)
Thermo Scientific™ Nunc Cryotubes and accessories are designed to work together and provide the best solutions for the most trusted sample storage applications.
  • Non-cytotoxic, non-mutatgenic raw materials and pyrogen-free IVD compatible tubes for added sample security
  • External or internally threaded options available
  • Certified SAL of 10-6 for the highest sterility assurance available
  • Suitable for storage from general cold storage refrigeration (+4°C) down to the vapor phase of liquid nitrogen
  • Bulk, uncapped, non-sterile versions available for less demanding applications
Specifications
Closure ColorOrange
GraduationsYes
StyleWriting Surface, A-R Base
AutoclavableNot Autoclavable
Bottom ShapeConical (Starfoot)
Capacity (Metric)1 mL
GraduatedYes
MaterialPolypropylene
Product TypeCryogenic Storage Tube
SterilitySterile
Thread StyleExternal
Unit SizeCase of 2000