Blocker™ Casein in PBS, 100 mL - FAQs
Blocker™ Casein in PBS, 100 mL - FAQs
View additional product information for Blocker™ Casein - FAQs (37582, 37583, 37528X3, 37532, 37528)
1 product FAQs found
What is the main ingredient in Blocker Casein (Cat. No. 37582, 37528, 37528X3)?
Blocker Casein (Cat. No. 37582, 37528, 37528X3) is a ready-to-use, 1% (w/v) solution of purified casein in phosphate-buffered saline (PBS). It is a stable, thimerosal-free formulation and is already at the optimal concentration for most applications. However, it can be diluted further if needed.
Find additional tips, troubleshooting help, and resources within our Protein Biology Support Centers .